missing translation for 'onlineSavingsMsg'
Learn More

Pentane, Technical, 95%, Spectrum™ Chemical

Catalog No. p-7165998
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612169 500 mL Amber Glass Bottle
18612168 4 L Amber Glass Bottle
18612166 20 L Steel Can
18612167 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612169 Supplier Spectrum Chemical Mfg Cor Supplier No. P1014500MLGL
Only null left

Chemical Identifiers

CAS 109-66-0

Specifications

CAS Max % 1
Boiling Point 36°C
Quantity 500 mL
Packaging Amber Glass Bottle
Percent Purity 0.95
Grade Technical
Chemical Name or Material Pentane

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.