missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentane, Technical, 95%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Boiling Point | 36°C |
| Quantity | 500 mL |
| Packaging | Amber Glass Bottle |
| Percent Purity | 0.95 |
| Grade | Technical |
| Chemical Name or Material | Pentane |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.