missing translation for 'onlineSavingsMsg'
Learn More

Pentane, Reagent grade, 98%, Honeywell Burdick & Jackson™

Catalog No. p-7120648
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044914 1 L Glass Bottle
60044915 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60044914 Supplier Honeywell Chemicals Supplier No. 1589411L
Only null left

Reagent Grade, 98%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -131°C
Color Colorless
Density 0.626g/mL at 25°C
Quantity 1 L
Explosion Limit 0.083
Flash Point −49°C (−56.2°F)
Ignition Point 500°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.358(lit.)
Vapor Pressure 26.85psi (55°C), 8.43psi (20°C)
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Beilstein 969132
Boiling Point 37°C
Formula Weight 72.15g/mol
Percent Purity 98%
Grade Reagent
Chemical Name or Material Pentane
Show More Show Less
Hazard Statement P210-P301 + P310-P331-P370 + P378-P403 + P235
Precautionary Statement H225-H304-H336-H411
DOTInformation Class 3 - PG 2 - Pentanes
EINECSNumber 203-692-4
RTECSNumber RZ9450000