missing translation for 'onlineSavingsMsg'
Learn More

Pentane (Pesticide), Fisher Chemical

Catalog No. P4004
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P4004 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P4004 Supplier Thermo Fisher Scientific Supplier No. FLP4004
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130°C
Density 0.626g/cm³
Quantity 4 L
Identification Pass Test
Packaging Amber Glass
Physical Form Liquid
Vapor Pressure 573mbar at 20°C
Linear Formula CH3(CH2)3CH3
Viscosity 0.25 mPa/s at 20°C
Boiling Point 36°C
Formula Weight 72.15
Grade Pesticide
Chemical Name or Material Pentane
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause skin, eye, and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:1
Flammability:4
Instability:0