missing translation for 'onlineSavingsMsg'
Learn More

Pentane, HPLC Grade, 99+% (n-Pentane, 95% min)

CAS: 109-66-0 | C5H12 | 72.15 g/mol

Supplier:  Thermo Scientific Chemicals H274270F

 View more versions of this product

Catalog No. AAH274270F


Only null left
Explore available promotions

Description

Description

n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Pentane is used in consumer merchandise such as spot lifters/cleaners and foaming shave gels and in commercial products such as a blowing agent for expanded polystyrene (foam) insulation. It may also be used as a solvent (in pure form) in laboratory, chemical analysis, and other settings where a highly volatile non-polar solvent is needed.

Solubility
Miscible with water.

Notes
Stable under normal ambient and anticipated storage and handling conditions of temperature. Avoidable conditions are heat, sparks, fire, and oxidizing agents.
Specifications

Specifications

Pentane
109-66-0
36°C
−49°C (−57°F)
1.358
MFCD00009498
14,7116
Miscible with water.
CCCCC
72.15
CHEBI:37830
≥99%
-130°C
0.626
2500 mL
Nearly Odorless
C5H12
UN1265
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
OFBQJSOFQDEBGM-UHFFFAOYSA-N
pentane
8003
72.15
HPLC

Chemical Identifiers

109-66-0
72.15
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
pentane
C5H12
MFCD00009498
n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
CHEBI:37830
CCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c

H225-H304-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PENTANES

EINECSNumber : 203-692-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only