missing translation for 'onlineSavingsMsg'
Learn More

Pentane (HPLC), Fisher Chemical™

Catalog No. p-216264
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass
Safe-Cote™ Amber Glass 
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P3991 1 L Amber Glass
P399SK1 1 L Safe-Cote™ Amber Glass 
P3994 4 L Amber Glass
P399SK4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. P3991 Supplier Thermo Fisher Scientific Supplier No. P399 1
Only null left

Submicron filtered

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130°C
Absorbance 0.014 AU max. at 254nm, 0.1 AU max. at 210nm, 0.3 AU max. at 200nm, 1.0 AU max. at 190nm
Density 0.626g/cm³
Quantity 1 L
Flash Point −49°C (−56.27deg;C)
Identification Pass Test
Packaging Amber Glass
Physical Form Liquid
Refractive Index 1.3540 to 1.3570 (at 25°C)
Residue after Evaporation 5ppm max.
Assay Percent Range >99%
Linear Formula CH3(CH2)3CH3
Vapor Pressure 573mbar at 20°C
Viscosity 0.25 mPa/s at 20°C
UN Number 1265
Water 0.02% max.
Boiling Point 36°C
Formula Weight 72.15
Percent Purity >99%
Grade Mobile phase for HPLC applications
Chemical Name or Material Pentane
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Extremely flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause skin, eye, and respiratory tract irritation. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:1
Flammability:4
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid