missing translation for 'onlineSavingsMsg'
Learn More

Pentane, CHROMASOLV™, for pesticide residue analysis, Solstice

Catalog No. 6001118
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001118 1 L
6001119 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001118 Supplier Solstice Supplier No. 344971L

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Density 0.626g/mL at 25°C
Quantity 1 L
Explosion Limit 0.083
Flash Point −49°C (−56.2°F)
Ignition Point 500°F
Linear Formula CH3(CH2)3CH3
UN Number UN1265
Beilstein 969132
Formula Weight 72.15g/mol
Percent Purity ≥99% (GC)
Grade CHROMASOLV
Chemical Name or Material Pentane
Show More Show Less
Hazard Statement P210-P261-P273-P301 + P310-P331
RTECSNumber RZ9450000