missing translation for 'onlineSavingsMsg'
Learn More

Pentane-1-sulfonic acid sodium salt, LiChropur™ EMD-Millipore

Catalog No. M1183040025
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1183040025 25 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1183040025 Supplier Technidata Supplier No. 1.18304.0025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ion pair reagents for analytical HPLC LiChropur

Chemical Identifiers

CAS 22767-49-3
Molecular Formula C5H11NaO3S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00007541,MFCD00149548
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
Synonym sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt
PubChem CID 23664617
IUPAC Name sodium pentane-1-sulfonate
SMILES [Na+].CCCCCS([O-])(=O)=O

Specifications

Density 390kg/m3
Packaging Glass
Quantity 25 g
Formula Weight 174.2
Percent Purity ≥99%
Grade Not Available
Physical Form Solid
Chemical Name or Material Pentane-1-sulfonic acid sodium salt
Recommended Storage Store at 5° to 30°C