missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylene Sulfide 98.0+%, TCI America™

Catalog No. P00445G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P00445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P00445G Supplier TCI America Supplier No. P00445G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1613-51-0
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.195
MDL Number MFCD00006662
InChI Key YPWFISCTZQNZAU-UHFFFAOYSA-N
Synonym pentamethylene sulfide, tetrahydrothiopyran, tetrahydro-2h-thiopyran, thiacyclohexane, 2h-thiopyran, tetrahydro, penthiophane, tetrahydrothiapyran, tetrahydrothiopyrane, thian, thian-4-yl
PubChem CID 15367
IUPAC Name thiane
SMILES C1CCSCC1

Specifications

Color Yellow
Boiling Point 140°C
Quantity 5 g
UN Number 1993
Formula Weight 102.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentamethylene Sulfide