missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentamethylene Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoate]dizinc(II) 97.0+%, TCI America™
[Reagent for application of the exciton chirality method]
Supplier: TCI America P1364100MG
Specifications
| Pentamethylene Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoate]dizinc(II) [Reagent for application | |
| Purple | |
| MFCD02093489 | |
| 1,5-Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoyloxy]pentane Dizinc(II), Pentamethylene Bis[4-(10,15,20-triphenylporphin-5-yl)benzoate]dizinc(II) | |
| C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=C(C=C8)C(=O)OCCCCCOC(=O)C9=CC=C(C=C9)C1=C2C=CC(=N2)C(=C2C=CC(=C(C5=NC(=C(C6=CC=C1[N-]6)C1=CC=CC=C1)C=C5)C1=CC=CC=C1)[N-]2)C1=CC=CC=C1)C=C4)C1=CC=CC=C1)[N-]3.[Zn+2].[Zn+2] | |
| 53384584 | |
| ≥97.0% (HPLC) |
| 210769-64-5 | |
| C95H64N8O4Zn2 | |
| 100 mg | |
| NBWTUFLOLJEGJZ-UHFFFAOYSA-L | |
| 1512.369 | |
| 1512.37 | |
| Crystalline Powder |
Chemical Identifiers
| 210769-64-5 | |
| 1512.369 | |
| NBWTUFLOLJEGJZ-UHFFFAOYSA-L | |
| 53384584 |
| C95H64N8O4Zn2 | |
| MFCD02093489 | |
| 1,5-Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoyloxy]pentane Dizinc(II), Pentamethylene Bis[4-(10,15,20-triphenylporphin-5-yl)benzoate]dizinc(II) | |
| C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=C(C=C8)C(=O)OCCCCCOC(=O)C9=CC=C(C=C9)C1=C2C=CC(=N2)C(=C2C=CC(=C(C5=NC(=C(C6=CC=C1[N-]6)C1=CC=CC=C1)C=C5)C1=CC=CC=C1)[N-]2)C1=CC=CC=C1)C=C4)C1=CC=CC=C1)[N-]3.[Zn+2].[Zn+2] |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator