missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylene Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoate]dizinc(II) 97.0+%, TCI America™

Catalog No. P136410MG
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P136410MG 10 mg
P1364100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P136410MG Supplier TCI America Supplier No. P136410MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[Reagent for application of the exciton chirality method]

Chemical Identifiers

CAS 210769-64-5
Molecular Formula C95H64N8O4Zn2
Molecular Weight (g/mol) 1512.369
MDL Number MFCD02093489
InChI Key NBWTUFLOLJEGJZ-UHFFFAOYSA-L
Synonym 1,5-Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoyloxy]pentane Dizinc(II), Pentamethylene Bis[4-(10,15,20-triphenylporphin-5-yl)benzoate]dizinc(II)
PubChem CID 53384584
SMILES C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=C(C=C8)C(=O)OCCCCCOC(=O)C9=CC=C(C=C9)C1=C2C=CC(=N2)C(=C2C=CC(=C(C5=NC(=C(C6=CC=C1[N-]6)C1=CC=CC=C1)C=C5)C1=CC=CC=C1)[N-]2)C1=CC=CC=C1)C=C4)C1=CC=CC=C1)[N-]3.[Zn+2].[Zn+2]

Specifications

Color Purple
Quantity 10 mg
Formula Weight 1512.37
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pentamethylene Bis[4-(10,15,20-triphenylporphyrin-5-yl)benzoate]dizinc(II) [Reagent for application
TSCA No
Recommended Storage Refrigerator