missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Pentamethylcyclopentadienyl)rhodium(III) Dichloride Dimer 96.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 618.07 |
| Percent Purity | ≥96.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (Pentamethylcyclopentadienyl)rhodium(III) Dichloride Dimer |
Safety and Handling
| TSCA | No |