missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluoropyridine 98.0+%, TCI America™
$123.03 - $391.44
Chemical Identifiers
| CAS | 700-16-3 |
|---|---|
| Molecular Formula | C5F5N |
| Molecular Weight (g/mol) | 169.054 |
| MDL Number | MFCD00006225 |
| InChI Key | XTGOWLIKIQLYRG-UHFFFAOYSA-N |
| Synonym | pentafluoropyridine, perfluoropyridine, pyridine, pentafluoro, pentafluorpyridin, pubchem9299, acmc-1bl6p, 2,3,4,5,6-pentafluoro-pyridine |
| PubChem CID | 69690 |
| IUPAC Name | 2,3,4,5,6-pentafluoropyridine |
| SMILES | C1(=C(C(=NC(=C1F)F)F)F)F |
Chemical Identifiers
| 700-16-3 | |
| 169.054 | |
| XTGOWLIKIQLYRG-UHFFFAOYSA-N | |
| 69690 | |
| C1(=C(C(=NC(=C1F)F)F)F)F |
| C5F5N | |
| MFCD00006225 | |
| pentafluoropyridine, perfluoropyridine, pyridine, pentafluoro, pentafluorpyridin, pubchem9299, acmc-1bl6p, 2,3,4,5,6-pentafluoro-pyridine | |
| 2,3,4,5,6-pentafluoropyridine |
Specifications
| 700-16-3 | |
| 83°C | |
| MFCD00006225 | |
| 1993 | |
| XTGOWLIKIQLYRG-UHFFFAOYSA-N | |
| 2,3,4,5,6-pentafluoropyridine | |
| 69690 | |
| ≥98.0% (GC) | |
| Pentafluoropyridine |
| Colorless | |
| C5F5N | |
| 5 g | |
| pentafluoropyridine, perfluoropyridine, pyridine, pentafluoro, pentafluorpyridin, pubchem9299, acmc-1bl6p, 2,3,4,5,6-pentafluoro-pyridine | |
| C1(=C(C(=NC(=C1F)F)F)F)F | |
| 169.054 | |
| 169.05 | |
| Liquid |
Safety and Handling
TSCA : No