missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropyridine 98.0+%, TCI America™

Catalog No. P09265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09265G 5 g
P092625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09265G Supplier TCI America Supplier No. P09265G

missing translation for 'validMainframeMsg'

CAS: 700-16-3 | C5F5N | 169.054 g/mol

Chemical Identifiers

CAS 700-16-3
Molecular Formula C5F5N
Molecular Weight (g/mol) 169.054
MDL Number MFCD00006225
InChI Key XTGOWLIKIQLYRG-UHFFFAOYSA-N
Synonym pentafluoropyridine, perfluoropyridine, pyridine, pentafluoro, pentafluorpyridin, pubchem9299, acmc-1bl6p, 2,3,4,5,6-pentafluoro-pyridine
PubChem CID 69690
IUPAC Name 2,3,4,5,6-pentafluoropyridine
SMILES C1(=C(C(=NC(=C1F)F)F)F)F

Specifications

Color Colorless
Boiling Point 83°C
Quantity 5 g
UN Number 1993
Formula Weight 169.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentafluoropyridine