missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropyridine, 99+%

CAS: 700-16-3 | C5F5N | 169.05 g/mol

$100.18 - $273.28

Chemical Identifiers

CAS 700-16-3
Molecular Formula C5F5N
Molecular Weight (g/mol) 169.05
MDL Number MFCD00006225
InChI Key XTGOWLIKIQLYRG-UHFFFAOYSA-N
Synonym pentafluoropyridine, perfluoropyridine, pyridine, pentafluoro, pentafluorpyridin, pubchem9299, acmc-1bl6p, 2,3,4,5,6-pentafluoro-pyridine
PubChem CID 69690
IUPAC Name 2,3,4,5,6-pentafluoropyridine
SMILES C1(=C(C(=NC(=C1F)F)F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169990050
SDS View Documents
Thermo Scientific Chemicals
169990050
5 g Glass bottle
Each for $100.18
Only null left
 
AC169990250
SDS View Documents
Thermo Scientific Chemicals
169990250
25 g Glass bottle
Each for $273.28
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

700-16-3
169.05
XTGOWLIKIQLYRG-UHFFFAOYSA-N
69690
C1(=C(C(=NC(=C1F)F)F)F)F
C5F5N
MFCD00006225
pentafluoropyridine, perfluoropyridine, pyridine, pentafluoro, pentafluorpyridin, pubchem9299, acmc-1bl6p, 2,3,4,5,6-pentafluoro-pyridine
2,3,4,5,6-pentafluoropyridine

Specifications

700-16-3
1.5400g/mL
23°C
99+%
C5F5N
(C5N)F5
5 g
pentafluoropyridine, perfluoropyridine, pyridine, pentafluoro, pentafluorpyridin, pubchem9299, acmc-1bl6p, 2,3,4,5,6-pentafluoro-pyridine
C1(=C(C(=NC(=C1F)F)F)F)F
169.05
169.05
Liquid
Colorless
84°C
Authentic
Glass bottle
1.3850 to 1.387
MFCD00006225
1.54
XTGOWLIKIQLYRG-UHFFFAOYSA-N
2,3,4,5,6-pentafluoropyridine
69690
99+%
Pentafluoropyridine, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 211-839-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only