missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropyridine, 99%

Catalog No. p-7024638
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1965606 5 g
AAA1965614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1965606 Supplier Thermo Scientific Chemicals Supplier No. A1965606

CAS: 700-16-3 | C5F5N | 169.054 g/mol

Pentafluoropyridine was used in the preparation of η2-C,C coordinated pentafluoropyridine complex. It was also used in the preparation of polyfluorinated coupling products via Pd(0)-catalyzed cross-coupling reaction with diarylzinc compounds. It was also used as derivatizing reagent in the sensitive GC-MS method for determination of the four endocrine disrupting chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluoropyridine was used in the preparation of η2-C,C coordinated pentafluoropyridine complex. It was also used in the preparation of polyfluorinated coupling products via Pd(0)-catalyzed cross-coupling reaction with diarylzinc compounds. It was also used as derivatizing reagent in the sensitive GC-MS method for determination of the four endocrine disrupting chemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents, alkali metals and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 700-16-3
Molecular Formula C5F5N
Molecular Weight (g/mol) 169.054
MDL Number MFCD00006225
InChI Key XTGOWLIKIQLYRG-UHFFFAOYSA-N
Synonym pentafluoropyridine, perfluoropyridine, pyridine, pentafluoro, pentafluorpyridin, pubchem9299, acmc-1bl6p, 2,3,4,5,6-pentafluoro-pyridine
PubChem CID 69690
IUPAC Name 2,3,4,5,6-pentafluoropyridine
SMILES C1(=C(C(=NC(=C1F)F)F)F)F

Specifications

Density 1.618
Boiling Point 83°C to 85°C
Flash Point 23°C (73°F)
Refractive Index 1.3865
Quantity 5 g
UN Number UN2924
Beilstein 1427064
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 169.05
Percent Purity 99%
Chemical Name or Material Pentafluoropyridine
Show More Show Less
Hazard Category H226-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 211-839-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only