missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Pentafluoropropionic anhydride

Catalog No. 25238725G
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25238725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25238725G Supplier Sigma Aldrich Supplier No. 25238725G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Pentafluoropropionic anhydride was used:
  • In preparation of halogenated derivatives of phenolic acids, required for determination of some biologically important acids in cerebrospinal fluid
  • As derivatization reagent in detection of 1,2,3,4-tetrahydroisoquinoline and 1-methyl-1,2,3,4-tetrahydroisoquinoline as endogenous amines from non-treated rat brain by GC-multiple ion detection
  • In simultaneous determination of the major metabolites of dopamine in rat striatum by gas-liquid chromatography

Specifications

CAS 356-42-3
Density 1.571 g/mL (at 25°C (literature))
Boiling Point 69°C to 70°C (735 mmHg)
Molecular Formula C6F10O3
Linear Formula (CF3CF2CO)2 O
MDL Number MFCD00000429
Quantity 25 g
Synonym PFPA; Perfluoropropionic anhydride
Molecular Weight (g/mol) 310.05
Percent Purity 99%