missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropropionic Acid 98.0+%, TCI America™

Catalog No. P112525G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P112525G 25 g
P1125100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P112525G Supplier TCI America Supplier No. P112525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 422-64-0
Molecular Formula C3HF5O2
Molecular Weight (g/mol) 164.03
MDL Number MFCD00004170
InChI Key LRMSQVBRUNSOJL-UHFFFAOYSA-N
Synonym pentafluoropropionic acid, perfluoropropionic acid, pentafluoropropanoic acid, propanoic acid, pentafluoro, propionic acid, pentafluoro, 2,2,3,3,3-pentafluoropropionic acid, ipc-pffa-3, perfluoropropanoic acid, propanoic acid, 2,2,3,3,3-pentafluoro, perfluoropropionicacid
PubChem CID 62356
IUPAC Name pentafluoropropanoic acid
SMILES OC(=O)C(F)(F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 98°C
Quantity 25 g
UN Number 3265
Formula Weight 164.03
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Pentafluoropropionic Acid
EINECSNumber (2)-1182
RTECSNumber UF6475000
TSCA Yes