missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluoropropionic acid, 97%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | Pentafluoropropionic acid |
| CAS | 422-64-0 |
| Densité | 1.5760g/mL |
| Point d’ébullition | 96°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 97% |
| Emballage | Plastic bottle |
| Formule moléculaire | C3AgF5O2 |
| Indice de réfraction | 1.2830 to 1.286 |
| Formule linéaire | CF3CF2CO2H |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement: Causes serious eye damage. Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 207-021-6 |
RUO – Research Use Only