missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropropionamide 98.0+%, TCI America™

Catalog No. P13165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13165G 5 g
P131625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13165G Supplier TCI America Supplier No. P13165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 354-76-7
Molecular Formula C3H2F5NO
Molecular Weight (g/mol) 163.05
MDL Number MFCD00039771
InChI Key KQTOYEUYHXUEDB-UHFFFAOYSA-N
Synonym pentafluoropropionamide, pentafluoropropanamide, propanamide, 2,2,3,3,3-pentafluoro, 2,2,3,3,3-pentafluoropropionamide, acmc-1cjql, pentafluoro propionic acid amide, 2,2,3,3,3-pentafluoropropionamide,, propanamide,2,2,3,3,3-pentafluoro
PubChem CID 67722
IUPAC Name pentafluoropropanamide
SMILES NC(=O)C(F)(F)C(F)(F)F

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 5 g
Formula Weight 163.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluoropropionamide