missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl pyridine-2-carboxylate, 95%, Thermo Scientific™

Catalog No. cc04026da
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC04026DA 1g Amber Glass Bottle
CC04026CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC04026DA Supplier Thermo Scientific Chemicals Supplier No. CC04026DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 188837-53-8
Molecular Formula C12H4F5NO2
Molecular Weight (g/mol) 289.16
MDL Number MFCD02916457
InChI Key KFJSLIBBYMXLTG-UHFFFAOYSA-N
Synonym pentafluorophenyl pyridine-2-carboxylate, pentafluorophenyl picolinate, 2-pyridinecarboxylicacid, 2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl pyridine-2-carboxylate, picolinic acid pentafluorophenyl ester, 2-pentafluorophenoxy carbonyl pyridine, 2,3,4,5,6-pentakis fluoranyl phenyl pyridine-2-carboxylate, 2-pyridinecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 2754830
IUPAC Name (2,3,4,5,6-pentafluorophenyl) pyridine-2-carboxylate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=CC=N2)C(F)=C1F

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 289.16
Percent Purity 95%
Chemical Name or Material Pentafluorophenyl pyridine-2-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.