missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC23826CB

 View more versions of this product

Catalog No. CC23826CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate
96.5
97%
C17H9F5N2O2
250mg
LGHZGIHYFYLEAC-UHFFFAOYSA-N
2,3,4,5,6-pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate
24229473
97%
915707-42-5
100.0
Amber Glass Bottle
MFCD09702352
pentafluorophenyl 4-1-methyl-1h-pyrazol-3-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-1-methylpyrazol-3-yl benzoate, pentafluorophenyl 4-1-methylpyrazol-3-yl benzoate, perfluorophenyl 4-1-methyl-1h-pyrazol-3-yl benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-1-methylpyrazol-3-yl benzoate, 4-1-methyl-3-pyrazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
CN1C=CC(=N1)C1=CC=C(C=C1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
368.26
368.26

Chemical Identifiers

915707-42-5
368.26
LGHZGIHYFYLEAC-UHFFFAOYSA-N
24229473
CN1C=CC(=N1)C1=CC=C(C=C1)C(=O)OC1=C(F)C(F)=C(F)C(F)=C1F
C17H9F5N2O2
MFCD09702352
pentafluorophenyl 4-1-methyl-1h-pyrazol-3-yl benzoate, 2,3,4,5,6-pentafluorophenyl 4-1-methylpyrazol-3-yl benzoate, pentafluorophenyl 4-1-methylpyrazol-3-yl benzoate, perfluorophenyl 4-1-methyl-1h-pyrazol-3-yl benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-1-methylpyrazol-3-yl benzoate, 4-1-methyl-3-pyrazolyl benzoic acid 2,3,4,5,6-pentafluorophenyl ester
2,3,4,5,6-pentafluorophenyl 4-(1-methyl-1H-pyrazol-3-yl)benzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions