missing translation for 'onlineSavingsMsg'
Learn More

Pentafluorophenyl 1-methyl-1H-indole-7-carboxylate, 97%, Thermo Scientific™

Catalog No. p-3586880
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC66926CB 250mg Amber Glass Bottle
CC66926DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC66926CB Supplier Thermo Scientific Chemicals Supplier No. CC66926CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 941716-96-7
Molecular Formula C16H8F5NO2
Molecular Weight (g/mol) 341.24
MDL Number MFCD09879962
InChI Key UEOKRSIJVAQHID-UHFFFAOYSA-N
Synonym pentafluorophenyl 1-methyl-1h-indole-7-carboxylate, pentafluorophenyl 1-methylindole-7-carboxylate, 2,3,4,5,6-pentakis fluoranyl phenyl 1-methylindole-7-carboxylate, 1-methyl-7-indolecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229744
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 1-methylindole-7-carboxylate
SMILES CN1C=CC2=CC=CC(C(=O)OC3=C(F)C(F)=C(F)C(F)=C3F)=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 341.23
Percent Purity 97%
Chemical Name or Material Pentafluorophenyl 1-methyl-1H-indole-7-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.