missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Pentafluoroethyl)trimethylsilane, 97%
Description
(Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.
Solubility
Miscible with tetrahydrofuran.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
(Pentafluoroethyl)trimethylsilane acts as a silicon-based fluorinating reagent for trifluoromethylation of carbonyls and ethylene diamines. It is also employed as a precursor for metal-mediated trifluoromethylation of aromatic, heteroaromatic and vinyl substrates.
Solubility
Miscible with tetrahydrofuran.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 1 g |
| UN Number | UN1993 |
| Solubility Information | Miscible with tetrahydrofuran. |
| Formula Weight | 192.21 |
| Percent Purity | 97% |
| Refractive Index | 1.327 |
| Sensitivity | Moisture sensitive |
| Chemical Name or Material | (Pentafluoroethyl)trimethylsilane |
Safety and Handling
| Hazard Category | H225-H315-H319-H335 |
| Hazard Statement | GHS H Statement H224-H315-H319-H335 Extremely flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only