missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoroethanesulfonamide 98.0+%, TCI America™

Numéro de catalogue. P23945G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P23945G 5 g
P23941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P23945G Fournisseur TCI America Code fournisseur P23945G
Il en reste null

Identifiants chimiques

CAS 78491-70-0
Molecular Formula C2H2F5NO2S
Molecular Weight (g/mol) 199.095
MDL Number MFCD19442187
InChI Key QDUKVMNUAQTMQW-UHFFFAOYSA-N
Synonym Perfluoroethanesulfonamide
PubChem CID 11252905
IUPAC Name 1,1,2,2,2-pentafluoroethanesulfonamide
SMILES C(C(F)(F)S(=O)(=O)N)(F)(F)F

Spécifications

Melting Point 80°C
Color White
Quantity 5 g
Formula Weight 199.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluoroethanesulfonamide