missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorobenzyl Methacrylate (stabilized with BHT) 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 266.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Pentafluorobenzyl Methacrylate (stabilized with BHT) |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |