missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluorobenzoic acid, 99%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Boiling Point | 220.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Linear Formula | C6F5CO2H |
| Quantity | 100g |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in hot water |
| Formula Weight | 212.07 |
| Percent Purity | 99% |
| Chemical Name or Material | Pentafluorobenzoic acid |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and kee |
| EINECSNumber | 210-026-6 |
RUO – Research Use Only