missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaethylenehexamine (so called), TCI America™
$48.46 - $132.35
Chemical Identifiers
| CAS | 4067-16-7 |
|---|---|
| Molecular Formula | C10H28N6 |
| Molecular Weight (g/mol) | 232.376 |
| MDL Number | MFCD00008167 |
| InChI Key | LSHROXHEILXKHM-UHFFFAOYSA-N |
| Synonym | pentaethylenehexamine, peha, 3,6,9,12-tetraazatetradecane-1,14-diamine, 3,6,9,12-tetraazatetradecamethylenediamine, unii-9k5vzu43le, ccris 6264, n1,n1'-ethane-1,2-diyl bis n2-2-aminoethyl ethane-1,2-diamine, 9k5vzu43le, n'-2-2-2-2-aminoethylamino ethylamino ethylamino ethyl ethane-1,2-diamine, pentaethylenexamine |
| PubChem CID | 19990 |
| IUPAC Name | N'-[2-[2-[2-(2-aminoethylamino)ethylamino]ethylamino]ethyl]ethane-1,2-diamine |
| SMILES | C(CNCCNCCNCCNCCN)N |
Chemical Identifiers
| 4067-16-7 | |
| 232.376 | |
| LSHROXHEILXKHM-UHFFFAOYSA-N | |
| 19990 | |
| C(CNCCNCCNCCNCCN)N |
| C10H28N6 | |
| MFCD00008167 | |
| pentaethylenehexamine, peha, 3,6,9,12-tetraazatetradecane-1,14-diamine, 3,6,9,12-tetraazatetradecamethylenediamine, unii-9k5vzu43le, ccris 6264, n1,n1'-ethane-1,2-diyl bis n2-2-aminoethyl ethane-1,2-diamine, 9k5vzu43le, n'-2-2-2-2-aminoethylamino ethylamino ethylamino ethyl ethane-1,2-diamine, pentaethylenexamine | |
| N'-[2-[2-[2-(2-aminoethylamino)ethylamino]ethylamino]ethyl]ethane-1,2-diamine |
Specifications
| 4067-16-7 | |
| MFCD00008167 | |
| 2735 | |
| LSHROXHEILXKHM-UHFFFAOYSA-N | |
| N'-[2-[2-[2-(2-aminoethylamino)ethylamino]ethylamino]ethyl]ethane-1,2-diamine | |
| 19990 | |
| Liquid |
| C10H28N6 | |
| 25 g | |
| pentaethylenehexamine, peha, 3,6,9,12-tetraazatetradecane-1,14-diamine, 3,6,9,12-tetraazatetradecamethylenediamine, unii-9k5vzu43le, ccris 6264, n1,n1'-ethane-1,2-diyl bis n2-2-aminoethyl ethane-1,2-diamine, 9k5vzu43le, n'-2-2-2-2-aminoethylamino ethylamino ethylamino ethyl ethane-1,2-diamine, pentaethylenexamine | |
| C(CNCCNCCNCCNCCN)N | |
| 232.376 | |
| 232.38 | |
| Pentaethylenehexamine (so called) |
Safety and Handling
EINECSNumber : (2)-0164
RTECSNumber : RZ2680000
TSCA : Yes