missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaethylenehexamine (so called), TCI America™
Specifications
Specifications
| Quantity | 25 g |
| UN Number | 2735 |
| Formula Weight | 232.38 |
| Physical Form | Liquid |
| Chemical Name or Material | Pentaethylenehexamine (so called) |
Safety and Handling
| EINECSNumber | (2)-0164 |
| RTECSNumber | RZ2680000 |
| TSCA | Yes |