Learn More
Pentaethylene glycol, 98+%
Used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors. | CAS: 4792-15-8 | C10H22O6 | 238.28 g/mol
Supplier: Thermo Scientific Chemicals A1013318
Description
Pentaethylene glycol is used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPentaethylene glycol is used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors.
Solubility
Miscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
Specifications
| Pentaethylene glycol | |
| Colorless | |
| 338°C to 340°C | |
| Glass bottle | |
| 1.462 | |
| 50 g | |
| 1.126 | |
| Miscible with water. | |
| C(COCCOCCOCCOCCO)O | |
| 238.28 | |
| CHEBI:39631 | |
| ≥98% |
| 4792-15-8 | |
| 1.126 | |
| >110°C (230°F) | |
| C10H22O6 | |
| MFCD00002878 | |
| 1635593 | |
| pentaethylene glycol, 3,6,9,12-tetraoxatetradecane-1,14-diol, pentaethyleneglycol, pentaglycol, ho ch2ch2o 5h, 3,6,9,12-tetraoxatetradocane-1,14-diol, 2-2-2-2-2-hydroxyethoxy ethoxy ethoxy ethoxy ethanol, dsstox_cid_7579, dsstox_rid_78510, dsstox_gsid_27579 | |
| JLFNLZLINWHATN-UHFFFAOYSA-N | |
| 2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol | |
| 62551 | |
| 238.28 | |
| Liquid |
Chemical Identifiers
| 4792-15-8 | |
| 238.28 | |
| JLFNLZLINWHATN-UHFFFAOYSA-N | |
| 62551 | |
| 2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethanol |
| C10H22O6 | |
| MFCD00002878 | |
| pentaethylene glycol, 3,6,9,12-tetraoxatetradecane-1,14-diol, pentaethyleneglycol, pentaglycol, ho ch2ch2o 5h, 3,6,9,12-tetraoxatetradocane-1,14-diol, 2-2-2-2-2-hydroxyethoxy ethoxy ethoxy ethoxy ethanol, dsstox_cid_7579, dsstox_rid_78510, dsstox_gsid_27579 | |
| CHEBI:39631 | |
| C(COCCOCCOCCOCCO)O |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 225-341-4
RTECSNumber : RZ2670000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only