missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol Tetrastearate (so called) 95.0+%, TCI America™

Catalog No. P073925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P073925G 25 g
P0739500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P073925G Supplier TCI America Supplier No. P073925G

Chemical Identifiers

CAS 115-83-3
Molecular Formula C77H148O8
Molecular Weight (g/mol) 1202.02
MDL Number MFCD00041922
InChI Key OCKWAZCWKSMKNC-UHFFFAOYSA-N
Synonym pentaerythrityl tetrastearate, pentaerythritol tetrastearate, unii-w9q3dzs0eg, stearic acid, neopentanetetrayl ester, w9q3dzs0eg, 3-octadecanoyloxy-2,2-bis octadecanoyloxymethyl propyl octadecanoate, octadecanoic acid, 2,2-bis 1-oxooctadecyl oxy methyl-1,3-propanediyl ester, pe tetrastearate, pentaerythrittetrastearat
PubChem CID 61037
IUPAC Name 3-(octadecanoyloxy)-2,2-bis[(octadecanoyloxy)methyl]propyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)(COC(=O)CCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCC

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 25 g
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pentaerythritol Tetrastearate (so called)
EINECSNumber (2)-0642
TSCA Yes