missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythritol Tetrakis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] 95.0+%, TCI America™
$48.46 - $203.17
Chemical Identifiers
| CAS | 6683-19-8 |
|---|---|
| Molecular Formula | C73H108O12 |
| Molecular Weight (g/mol) | 1177.66 |
| MDL Number | MFCD00059345 |
| InChI Key | BGYHLZZASRKEJE-UHFFFAOYSA-N |
| Synonym | Tetrakis[methylene-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]methane |
| PubChem CID | 64819 |
| IUPAC Name | 3-{[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]oxy}-2,2-bis({[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]oxy}methyl)propyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate |
| SMILES | CC(C)(C)C1=CC(CCC(=O)OCC(COC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)(COC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)COC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C |
Chemical Identifiers
| 6683-19-8 | |
| 1177.66 | |
| BGYHLZZASRKEJE-UHFFFAOYSA-N | |
| 64819 | |
| CC(C)(C)C1=CC(CCC(=O)OCC(COC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)(COC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)COC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C |
| C73H108O12 | |
| MFCD00059345 | |
| Tetrakis[methylene-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]methane | |
| 3-{[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]oxy}-2,2-bis({[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]oxy}methyl)propyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate |
Specifications
| 6683-19-8 | |
| White | |
| MFCD00059345 | |
| Tetrakis[methylene-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]methane | |
| CC(C)(C)C1=CC(CCC(=O)OCC(COC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)(COC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)COC(=O)CCC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C | |
| 1177.66 | |
| 1177.66 | |
| Crystalline Powder |
| 115°C | |
| C73H108O12 | |
| 25 g | |
| BGYHLZZASRKEJE-UHFFFAOYSA-N | |
| 3-{[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]oxy}-2,2-bis({[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl]oxy}methyl)propyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate | |
| 64819 | |
| ≥95.0% (HPLC) | |
| Pentaerythritol Tetrakis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] |
Safety and Handling
EINECSNumber : (3)-1693
RTECSNumber : DA8340900
TSCA : Yes