missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythritol Tetraacrylate (stabilized with MEHQ) 80.0+%, TCI America™
$67.11 - $264.70
Chemical Identifiers
| CAS | 4986-89-4 |
|---|---|
| Molecular Formula | C17H20O8 |
| Molecular Weight (g/mol) | 352.34 |
| MDL Number | MFCD00015334 |
| InChI Key | KNSXNCFKSZZHEA-UHFFFAOYSA-N |
| PubChem CID | 62556 |
| IUPAC Name | 3-(prop-2-enoyloxy)-2,2-bis[(prop-2-enoyloxy)methyl]propyl prop-2-enoate |
| SMILES | C=CC(=O)OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C |
Chemical Identifiers
| 4986-89-4 | |
| 352.34 | |
| KNSXNCFKSZZHEA-UHFFFAOYSA-N | |
| 3-(prop-2-enoyloxy)-2,2-bis[(prop-2-enoyloxy)methyl]propyl prop-2-enoate |
| C17H20O8 | |
| MFCD00015334 | |
| 62556 | |
| C=CC(=O)OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C |
Specifications
| 4986-89-4 | |
| C17H20O8 | |
| 25 g | |
| C=CC(=O)OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C | |
| 352.34 | |
| 352.34 | |
| Liquid |
| Yellow | |
| MFCD00015334 | |
| KNSXNCFKSZZHEA-UHFFFAOYSA-N | |
| 3-(prop-2-enoyloxy)-2,2-bis[(prop-2-enoyloxy)methyl]propyl prop-2-enoate | |
| 62556 | |
| ≥80.0% (GC) | |
| Pentaerythritol Tetraacrylate (stabilized with MEHQ) |
Safety and Handling
EINECSNumber : (2)-0958&(2)-2578
TSCA : Yes