missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentaerythritol Tetraacrylate (stabilized with MEHQ) 80.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 500 g |
| Formula Weight | 352.34 |
| Percent Purity | ≥80.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Pentaerythritol Tetraacrylate (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (2)-0958&(2)-2578 |
| TSCA | Yes |