missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol Tetraacrylate (stabilized with MEHQ) 80.0+%, TCI America™

Catalog No. P208425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P208425G 25 g
P2084500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P208425G Supplier TCI America Supplier No. P208425G
Only null left

Chemical Identifiers

CAS 4986-89-4
Molecular Formula C17H20O8
Molecular Weight (g/mol) 352.34
MDL Number MFCD00015334
InChI Key KNSXNCFKSZZHEA-UHFFFAOYSA-N
PubChem CID 62556
IUPAC Name 3-(prop-2-enoyloxy)-2,2-bis[(prop-2-enoyloxy)methyl]propyl prop-2-enoate
SMILES C=CC(=O)OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 352.34
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentaerythritol Tetraacrylate (stabilized with MEHQ)
EINECSNumber (2)-0958&(2)-2578
TSCA Yes