missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol Tetra(3-mercaptopropionate) 90.0+%, TCI America™

Catalog No. p-7134828
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P088625ML 25 mL
P0886500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P088625ML Supplier TCI America Supplier No. P088625ML

Chemical Identifiers

CAS 7575-23-7
Molecular Formula C17H28O8S4
Molecular Weight (g/mol) 488.64
MDL Number MFCD00022104
InChI Key JOBBTVPTPXRUBP-UHFFFAOYSA-N
PubChem CID 82056
IUPAC Name 3-[(3-sulfanylpropanoyl)oxy]-2,2-bis({[(3-sulfanylpropanoyl)oxy]methyl})propyl 3-sulfanylpropanoate
SMILES SCCC(=O)OCC(COC(=O)CCS)(COC(=O)CCS)COC(=O)CCS

Specifications

Color Colorless
Boiling Point 275°C
Quantity 25 mL
UN Number 3082
Formula Weight 488.64
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material Pentaerythritol Tetra(3-mercaptopropionate)
EINECSNumber (2)-3330
TSCA Yes