missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol 98.0+%, TCI America™

Catalog No. P003925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P003925G 25 g
P0039500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P003925G Supplier TCI America Supplier No. P003925G

Chemical Identifiers

CAS 115-77-5
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.147
MDL Number MFCD00004692
InChI Key WXZMFSXDPGVJKK-UHFFFAOYSA-N
Synonym pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek
PubChem CID 8285
IUPAC Name 2,2-bis(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)CO)O

Specifications

Melting Point 250°C
Color White
Boiling Point 276°C
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentaerythritol
EINECSNumber (2)-0248
RTECSNumber RZ2490000
TSCA Yes