missing translation for 'onlineSavingsMsg'
Learn More

Pentaerythritol, 98+%

Catalog No. AAA1052030
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1052030 250 g
AAA105200B 1000 g
AAA105200I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1052030 Supplier Thermo Scientific Chemicals Supplier No. A1052030
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 115-77-5 | C5H12O4 | 136.147 g/mol

Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.

Solubility
Soluble in Water: 0.1 g/mL

Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents, acid chlorides, acid anhydrides.

Chemical Identifiers

CAS 115-77-5
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.147
MDL Number MFCD00004692
InChI Key WXZMFSXDPGVJKK-UHFFFAOYSA-N
Synonym pentaerythritol, pentek, 2,2-bis hydroxymethyl propane-1,3-diol, tetramethylolmethane, monopentaerythritol, auxinutril, maxinutril, monopentek, pentaerythrite, penetek
PubChem CID 8285
IUPAC Name 2,2-bis(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)CO)O

Specifications

Melting Point 260°C to 264°C
Density 1.396
Boiling Point 276°C (30 mmHg)
Flash Point 240°C (464°F)
Quantity 250 g
Beilstein 1679274
Sensitivity Hygroscopic
Merck Index 14,7111
Solubility Information Soluble in Water: 0.1g/mL
Formula Weight 136.15
Percent Purity ≥98%
Chemical Name or Material Pentaerythritol
Show More Show Less
Hazard Category H500
EINECSNumber 204-104-9
RTECSNumber RZ2490000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only