missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentadecyl Acetate 95.0+%, TCI America™
Supplier: TCI America P170025G
Specifications
| Pentadecyl Acetate | |
| 15°C | |
| 230°C | |
| MFCD00056204 | |
| Acetic Acid Pentadecyl Ester | |
| CCCCCCCCCCCCCCCOC(=O)C | |
| 270.457 | |
| 270.46 | |
| Liquid |
| 629-58-3 | |
| Colorless | |
| C17H34O2 | |
| 25 g | |
| AAAIZLQILBFRTJ-UHFFFAOYSA-N | |
| pentadecyl acetate | |
| 522077 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 629-58-3 | |
| 270.457 | |
| AAAIZLQILBFRTJ-UHFFFAOYSA-N | |
| 522077 | |
| CCCCCCCCCCCCCCCOC(=O)C |
| C17H34O2 | |
| MFCD00056204 | |
| Acetic Acid Pentadecyl Ester | |
| pentadecyl acetate |
Safety and Handling
EINECSNumber : (2)-0735
TSCA : No