missing translation for 'onlineSavingsMsg'
Learn More

Pentadecanolide, 98%

Catalog No. AAA1134122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1134122 100 g
AAA1134114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1134122 Supplier Thermo Scientific Chemicals Supplier No. A1134122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-02-5 | C15H28O2 | 240.387 g/mol

Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentadecanolide is a monomer used to make PGA-co-pentadecalactone polymers to be used in the development of directly compressed prolonged release tablets of slightly soluble drugs such as ketoprofen. Pentadecanolide may be used for the synthesis of poly(pentadecanolide) (PPDL) by Novozyme 435 catalyzed ring-opening polymerization. It may be employed as starting reagent for the synthesis of terminally-branched iso-fatty acids (iso-C15-C17).

Solubility
Soluble in alcohol, dipropylene glycol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 106-02-5
Molecular Formula C15H28O2
Molecular Weight (g/mol) 240.387
MDL Number MFCD00039667
InChI Key FKUPPRZPSYCDRS-UHFFFAOYSA-N
Synonym cyclopentadecanolide, exaltolide, pentadecanolide, 15-pentadecanolide, pentalide, thibetolide, pentadecan-15-olide, pentadecalactone, muskalactone, muskolactone
PubChem CID 235414
IUPAC Name oxacyclohexadecan-2-one
SMILES C1CCCCCCCOC(=O)CCCCCC1

Specifications

Melting Point 33°C to 37°C
Density 0.918
Boiling Point 137°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.47
Quantity 100 g
Beilstein 143710
Merck Index 14,3910
Solubility Information Soluble in alcohol,dipropylene glycol. Insoluble in water.
Formula Weight 240.39
Percent Purity 98%
Chemical Name or Material Pentadecanolide
Show More Show Less
Hazard Category H317
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 203-354-6
RTECSNumber RN9775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only