missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentadecanedioic Acid 97.0+%, TCI America™
$55.92 - $333.67
Chemical Identifiers
| CAS | 1460-18-0 |
|---|---|
| Molecular Formula | C15H28O4 |
| Molecular Weight (g/mol) | 272.39 |
| MDL Number | MFCD00039534 |
| InChI Key | BTZVDPWKGXMQFW-UHFFFAOYSA-N |
| Synonym | 1,15-pentadecanedioic acid, pubchem18101, 1,15-pentadecanoic acid, 1,15-pentadecanedioicacid, acmc-1c0t0, ksc180a4d |
| PubChem CID | 160576 |
| IUPAC Name | pentadecanedioic acid |
| SMILES | OC(=O)CCCCCCCCCCCCCC(O)=O |
Chemical Identifiers
| 1460-18-0 | |
| 272.39 | |
| BTZVDPWKGXMQFW-UHFFFAOYSA-N | |
| 160576 | |
| OC(=O)CCCCCCCCCCCCCC(O)=O |
| C15H28O4 | |
| MFCD00039534 | |
| 1,15-pentadecanedioic acid, pubchem18101, 1,15-pentadecanoic acid, 1,15-pentadecanedioicacid, acmc-1c0t0, ksc180a4d | |
| pentadecanedioic acid |
Specifications
| 1460-18-0 | |
| White-Yellow | |
| C15H28O4 | |
| 1 g | |
| BTZVDPWKGXMQFW-UHFFFAOYSA-N | |
| pentadecanedioic acid | |
| 160576 | |
| ≥97.0% (GC,T) | |
| Pentadecanedioic Acid |
| 114°C | |
| 175°C | |
| MFCD00039534 | |
| 1,15-pentadecanedioic acid, pubchem18101, 1,15-pentadecanoic acid, 1,15-pentadecanedioicacid, acmc-1c0t0, ksc180a4d | |
| OC(=O)CCCCCCCCCCCCCC(O)=O | |
| 272.39 | |
| 272.39 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-3488
TSCA : Yes