missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Pentadecane

Catalog No. P3406500G
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P3406500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P3406500G Supplier Sigma Aldrich Supplier No. P3406500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentadecane

Specifications

CAS 629-62-9
Melting Point 8°C to 10°C (lit.)
Density 0.769 g/mL (at 25°C (literature))
Boiling Point 270°C (lit.)
Molecular Formula C15H32
Refractive Index n20/D 1.431 (literature)
Linear Formula CH3(CH2)13 CH3
MDL Number MFCD00008990
Quantity 500 g
Synonym n-Pentadecane
Molecular Weight (g/mol) 212.41
Percent Purity ≥99%
Show More Show Less