missing translation for 'onlineSavingsMsg'
Learn More

Pentadecafluoroheptyl Bromide 98.0+%, TCI America™

Catalog No. P17535G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P17535G Supplier TCI America Supplier No. P17535G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 375-88-2
Molecular Formula C7BrF15
Molecular Weight (g/mol) 448.96
MDL Number MFCD00013570
InChI Key VPQQZKWYZYVTMU-UHFFFAOYSA-N
Synonym 1-bromoperfluoroheptane, perfluoroheptyl bromide, 1-bromopentadecafluoroheptane, perfluoro-n-heptyl bromide, pentadecafluoroheptyl bromide, heptane, 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoro, acmc-1ad35, 1-bromopentadecafluoroheptane; perfluoroheptyl bromide, 1-bromanyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecakis fluoranyl heptane
PubChem CID 67819
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecafluoroheptane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Specifications

Color Yellow
Boiling Point 118°C
Quantity 5 g
Formula Weight 448.96
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentadecafluoroheptyl Bromide