missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentacosanoic Acid 96.0+%, TCI America™
$321.51 - $962.59
Chemical Identifiers
| CAS | 506-38-7 |
|---|---|
| Molecular Formula | C25H50O2 |
| Molecular Weight (g/mol) | 382.67 |
| MDL Number | MFCD00020551 |
| InChI Key | MWMPEAHGUXCSMY-UHFFFAOYSA-N |
| PubChem CID | 10468 |
| ChEBI | CHEBI:39420 |
| IUPAC Name | pentacosanoic acid |
| SMILES | CCCCCCCCCCCCCCCCCCCCCCCCC(O)=O |
Chemical Identifiers
| 506-38-7 | |
| 382.67 | |
| MWMPEAHGUXCSMY-UHFFFAOYSA-N | |
| CHEBI:39420 | |
| CCCCCCCCCCCCCCCCCCCCCCCCC(O)=O |
| C25H50O2 | |
| MFCD00020551 | |
| 10468 | |
| pentacosanoic acid |
Specifications
| 506-38-7 | |
| White-Yellow | |
| MFCD00020551 | |
| MWMPEAHGUXCSMY-UHFFFAOYSA-N | |
| pentacosanoic acid | |
| 10468 | |
| 382.67 | |
| Crystalline Powder |
| 83°C | |
| C25H50O2 | |
| 1 g | |
| CCCCCCCCCCCCCCCCCCCCCCCCC(O)=O | |
| 382.67 | |
| CHEBI:39420 | |
| ≥96.0% (GC) | |
| Pentacosanoic Acid |
Safety and Handling
EINECSNumber : (2)-0608
TSCA : No