missing translation for 'onlineSavingsMsg'
Learn More

Pentacosane 97.0+%, TCI America™

Catalog No. P01395G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P01395G 5 g
P01391G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P01395G Supplier TCI America Supplier No. P01395G

Chemical Identifiers

CAS 629-99-2
Molecular Formula C25H52
Molecular Weight (g/mol) 352.691
MDL Number MFCD00009353
InChI Key YKNWIILGEFFOPE-UHFFFAOYSA-N
Synonym n-pentacosane, unii-bon9h94y8v, bon9h94y8v, pentacosane, analytical standard, pentacosane, tetracosane, methyl, n-pentacosane 1g, pentacosane standardmaterialforgc, ch3-ch2 23-ch3, solution, 10,000 mg/l, 1 ml rm, iso guide 34
PubChem CID 12406
ChEBI CHEBI:32938
IUPAC Name pentacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 54°C
Color White
Boiling Point 254°C
Quantity 5 g
Formula Weight 352.69
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pentacosane
EINECSNumber (2)-0010
TSCA Yes