missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentachloroaniline 97.0+%, TCI America™
$240.46 - $961.86
Chemical Identifiers
| CAS | 527-20-8 |
|---|---|
| Molecular Formula | C6H2Cl5N |
| Molecular Weight (g/mol) | 265.339 |
| MDL Number | MFCD00014769 |
| InChI Key | KHCZSJXTDDHLGJ-UHFFFAOYSA-N |
| Synonym | pentachloroaniline, pentachloroaminobenzene, benzenamine, 2,3,4,5,6-pentachloro, pca van, unii-uw5qvl647i, 2,3,4,5,6-pentachlorobenzenamine, aniline, 2,3,4,5,6-pentachloro, uw5qvl647i, dsstox_cid_17584, dsstox_rid_79345 |
| PubChem CID | 10693 |
| IUPAC Name | 2,3,4,5,6-pentachloroaniline |
| SMILES | C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)N |
Chemical Identifiers
| 527-20-8 | |
| 265.339 | |
| KHCZSJXTDDHLGJ-UHFFFAOYSA-N | |
| 10693 | |
| C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)N |
| C6H2Cl5N | |
| MFCD00014769 | |
| pentachloroaniline, pentachloroaminobenzene, benzenamine, 2,3,4,5,6-pentachloro, pca van, unii-uw5qvl647i, 2,3,4,5,6-pentachlorobenzenamine, aniline, 2,3,4,5,6-pentachloro, uw5qvl647i, dsstox_cid_17584, dsstox_rid_79345 | |
| 2,3,4,5,6-pentachloroaniline |
Specifications
| 233°C | |
| White-Yellow | |
| Crystal-Powder at 20°C | |
| MFCD00014769 | |
| pentachloroaniline, pentachloroaminobenzene, benzenamine, 2,3,4,5,6-pentachloro, pca van, unii-uw5qvl647i, 2,3,4,5,6-pentachlorobenzenamine, aniline, 2,3,4,5,6-pentachloro, uw5qvl647i, dsstox_cid_17584, dsstox_rid_79345 | |
| C1(=C(C(=C(C(=C1Cl)Cl)Cl)Cl)Cl)N | |
| 265.339 | |
| 265.34 | |
| Pentachloroaniline |
| 527-20-8 | |
| 1 g | |
| C6H2Cl5N | |
| 2811 | |
| KHCZSJXTDDHLGJ-UHFFFAOYSA-N | |
| 2,3,4,5,6-pentachloroaniline | |
| 10693 | |
| ≥97.0% (GC) |
Safety and Handling
RTECSNumber : BY7910000
TSCA : No