missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentachloroaniline 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 233°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Physical Form | Crystal-Powder at 20°C |
| UN Number | 2811 |
| Formula Weight | 265.34 |
| Percent Purity | ≥97.0% (GC) |
| Chemical Name or Material | Pentachloroaniline |
Safety and Handling
| RTECSNumber | BY7910000 |
| TSCA | No |