missing translation for 'onlineSavingsMsg'
Learn More

Penta-O-acetyl-beta-D-glucopyranose 99.0+%, TCI America™

Catalog No. p-7134382
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0028100G 100 g
P0028500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0028100G Supplier TCI America Supplier No. P0028100G

Chemical Identifiers

CAS 604-69-3
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.341
MDL Number MFCD00006597
InChI Key LPTITAGPBXDDGR-IBEHDNSVSA-N
Synonym beta-d-glucose pentaacetate, penta-o-acetyl-beta-d-glucopyranose, unii-v833p95coc, 1,2,3,4,6-penta-o-acetyl-beta-d-glucopyranose, 1,2,3,4,6-penta-o-acetyl-b-d-glucopyranose, beta-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6s-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, 2s,3r,4s,5r,6r-6-acetoxymethyl tetrahydro-2h-pyran-2,3,4,5-tetrayl tetraacetate, glucopyranose, pentaacetate, .beta.-d, penta-o-acetyl-β
PubChem CID 2724702
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5,6-tetraacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 132°C
Color White
Quantity 100 g
Formula Weight 390.34
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Penta-O-acetyl-beta-D-glucopyranose
EINECSNumber (8)-0053
TSCA No