missing translation for 'onlineSavingsMsg'
Learn More

Penta-O-acetyl-alpha-D-glucopyranose 97.0+%, TCI America™

Catalog No. p-7130197
Change view
Click to view available options
Quantity:
25 g
250 g
Catalog No. Quantity
G022525G 25 g
G0225250G 250 g
2 options

Catalog No. G022525G

Supplier: TCI America G022525G

Only null left

Chemical Identifiers

CAS 604-68-2
Molecular Formula C16H22O11
Molecular Weight (g/mol) 390.34
MDL Number MFCD00064071,MFCD00064081
InChI Key LPTITAGPBXDDGR-UHFFFAOYNA-N
Synonym alpha-d-glucose pentaacetate, a-d-glucopyranose pentaacetate, pentaacetyl-alpha-d-glucose, penta-o-acetyl-alpha-d-glucopyranose, unii-1cs424ns93, 1,2,3,4,6-penta-o-acetyl-alpha-d-glucopyranose, alpha-d-glucopyranose pentaacetate, 2r,3r,4s,5r,6r-3,4,5,6-tetrakis acetyloxy oxan-2-yl methyl acetate, glucopyranose, pentaacetate, .alpha.-d, a-d-glucose pentaacetate
PubChem CID 2723636
IUPAC Name [3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 113°C
Color White
Quantity 25 g
Formula Weight 390.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Penta-O-acetyl-alpha-D-glucopyranose
EINECSNumber (8)-0053
TSCA Yes