Learn More
Gibco™ Penicillin Streptomycin GlutaMAX™ Supplement (100X)
Description
Gibco™ Penicillin Streptomycin GlutaMAX Supplement is a form of GlutaMAX Supplement that contains the antibiotics penicillin (10,000 units) and streptomycin (10,000 μg), along with 200 mM L-alanyl-L-glutamine dipeptide in a citrate buffer to maintain penicillin potency. GlutaMAX Supplement is an alternative to L-glutamine, with increased stability that improves cell health.
Compared to L-glutamine, GlutaMAX Supplement:
- Minimizes toxic ammonia build-up
- Improves cell viability and growth
Minimizes toxic ammonia build-up
Unlike L-glutamine, GlutaMAX Supplement does not spontaneously break down to form ammonia. Instead, cells cleave the dipeptide bond to release L-glutamine as needed. This system prevents waste build-up and maintains a fresh supply of L-glutamine during long-term culture.
Improves cell viability and growth
Cultures with lower levels of ammonia show improved cell viability. In addition, culture conditions are more controlled due to the consistent concentration of GlutaMAX Supplement that is not possible with L-glutamine.
About penicillin and streptomycin
The antibiotics penicillin and streptomycin are used to prevent bacterial contamination in cell cultures due to their effective combined action against gram-positive and gram-negative bacteria. Penicillin acts by interfering directly with the turnover of the bacteria cell wall and indirectly by triggering the release of enzymes that further alter the cell wall. Streptomycin acts by binding to the 30S subunit of the bacterial ribosome, leading to the inhibition of protein synthesis and death in susceptible bacteria.
Specifications
Specifications
| Concentration | 100X |
| Form | Clear, colorless to slight yellow solution |
| Product Type | Antibiotic |
| Sterility | Sterile-filtered |
| Sterilization Method | 0.2 μm Sterile-filtered |
| With Additives | GlutaMAX |
| Without Additives | None |
| Formulation | Liquid, citrate buffer |
| Osmolality | 330 to 370 mOsm/kg |
| pH | 4.7 to 6.2 |
| Show More |
Frequently Asked Questions (FAQs)
Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) contains the antibiotics penicillin (10,000 units) and streptomycin (10,000 µg), along with 200 mM L-alanyl-L-glutamine dipeptide, GlutaMAX supplement, in a citrate buffer to maintain penicillin potency.
Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) is an alternative to Penicillin-Streptomycin-Glutamine (100X) (Cat. No. 10378016), with increased stability that improves cell health. In aqueous solutions such as cell culture media, L-glutamine spontaneously degrades, generating toxic ammonia and pyrrolidine carboxylic acid as byproducts. GlutaMAX supplement is a dipeptide, L-alanyl-L-glutamine, which is more stable in cell culture media.
Yes, the product will still work. The bottle can be stored at 4 degrees C overnight to finish thawing and then aliquoted the next day for storage at -20 degrees C. Alternatively, the bottle can be stored at -20 degrees C until it is needed.
Yes, Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) may be stored at -80 degrees C.
Medium changes should be performed at least once every three days as penicillin and streptomycin are stable for three days at 37 degrees C.
For Research Use Only. Not for use in diagnostic procedures.